Invitrogen
Manufacturer Code:PA555969
Catalog # PIPA555969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC12A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na-Cl cotransporter; Na-Cl symporter; NaCl electroneutral thiazide-sensitive cotransporter; NCC; solute carrier family 12 (sodium/chloride transporter), member 3; Solute carrier family 12 (sodium/chloride transporters) member 3 NCCT TSC; Solute carrier family 12 member 3; thiazide-sensitive Na-Cl cotransporter; Thiazide-sensitive sodium-chloride cotransporter
Gene Aliases: NCC; NCCT; SLC12A3; TSC
UniProt ID: (Human) P55017
Entrez Gene ID: (Human) 6559
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.