Invitrogen
Manufacturer Code:PA542408
Catalog # PIPA542408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC12A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na-Cl cotransporter; Na-Cl symporter; NaCl electroneutral thiazide-sensitive cotransporter; NCC; NCCT; sodium chloride cotransporter; solute carrier family 12 (sodium/chloride transporter), member 3; Solute carrier family 12 (sodium/chloride transporters) member 3 NCCT TSC; solute carrier family 12 (sodium/chloride transporters), member 3; Solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; thiazide-sensitive sodium chloride cotransporter; Thiazide-sensitive sodium-chloride cotransporter
Gene Aliases: AI035291; NCC; NCCT; si:ch211-122c9.4; SLC12A3; TSC; wu:fj83c03
UniProt ID: (Human) P55017, (Rat) P55018, (Mouse) P59158
Entrez Gene ID: (Human) 6559, (Bovine) 404171, (Rabbit) 100009348, (Horse) 100051953, (Dog) 478118, (Rat) 54300, (Mouse) 20497, (Guinea pig) 100726300, (Zebrafish) 565439
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.