Invitrogen
Manufacturer Code:PA580003
Catalog # PIPA580003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human SLC12A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BSC1; bumetanide-sensitive cotransporter type 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; Kidney-specific Na-K-Cl symporter; Na-K-2Cl cotransporter; NKCC2A variant A; solute carrier family 12 (sodium/potassium/chloride transporter), member 1; solute carrier family 12 (sodium/potassium/chloride transporters), member 1; Solute carrier family 12 member 1; Solute carrier family 12 member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); Solute carrier family 12 member 1 Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2 Kidney-specific Na-K-Cl symporter SLC12A1 NKCC2; Solute carrier family 12, member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter)
Gene Aliases: AI788571; BSC1; D630042G03Rik; mBSC1; NKCC2; SLC12A1; urehr3
UniProt ID: (Human) Q13621, (Mouse) P55014
Entrez Gene ID: (Human) 6557, (Mouse) 20495, (Rat) 25065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.