Abnova
Manufacturer Code:PAB5884
Catalog # 89-131-975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Slc11a2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc11a2.
For Research Use Only
Protein Aliases: DCT1; Divalent cation transporter 1; Divalent metal transporter 1; DMT-1; DMT1; microcytic anemia, viable anaemia; mk; Natural resistance-associated macrophage protein 2; NRAMP 2; Nramp2; Solute carrier family 11 member 2; van
Gene Aliases: DCT1; DMT1; mk; Nramp2; Slc11a2; van
UniProt ID: (Mouse) P49282
Entrez Gene ID: (Mouse) 18174
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.