Abnova
Manufacturer Code:PAB27487
Catalog # 89-128-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC10A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC10A5.
For Research Use Only
Protein Aliases: Na(+)/bile acid cotransporter 5; Sodium/bile acid cotransporter 5; solute carrier family 10 (sodium/bile acid cotransporter family), member 5; Solute carrier family 10 member 5; solute carrier family 10, member 5
Gene Aliases: P5; SLC10A5
UniProt ID: (Human) Q5PT55
Entrez Gene ID: (Human) 347051
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.