Invitrogen
Manufacturer Code:PA523755
Catalog # PIPA523755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 370-398 amino acids from the C-terminal region of human SLC10A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bile acid transporter SLC10A4; Na(+)/bile acid cotransporter 4; P4 Na(+)/bile acid cotransporter 4 bile acid transporter SLC10A4 sodium/bile acid cotransporter 4 solute carrier family 10 member 4; Sodium/bile acid cotransporter 4; solute carrier family 10 (sodium/bile acid cotransporter family), member 4; Solute carrier family 10 member 4; solute carrier family 10, member 4
Gene Aliases: E130304D01; P4; SLC10A4
UniProt ID: (Human) Q96EP9, (Mouse) Q3UEZ8
Entrez Gene ID: (Human) 201780, (Mouse) 231290
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.