Novus Biologicals
Manufacturer Code:NBP15940120UL
Catalog # NBP1594020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC10A4(solute carrier family 10 (sodium/bile acid cotransporter family) member 4) The peptide sequence was selected from the N terminal of SLC10A4. Peptide sequence DGNDNVTLLFAPLLRDNYTLAPNASSLGPGTDLALA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bile acid transporter SLC10A4; bile acid transporter SLC10A4 MGC29802 Na(+)/bile acid cotransporter 4 P4 sodium/bile acid cotransporter 4 solute carrier family 10 (sodium/bile acid cotransporter family) member 4 Solute carrier family 10 member 4; Na(+)/bile acid cotransporter 4; Sodium/bile acid cotransporter 4; solute carrier family 10 (sodium/bile acid cotransporter family), member 4; Solute carrier family 10 member 4; solute carrier family 10, member 4
Gene Aliases: P4; SLC10A4
UniProt ID: (Human) Q96EP9
Entrez Gene ID: (Human) 201780
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.