Proteintech
Manufacturer Code:199091AP150UL
Catalog # 50-173-2595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC10A3 Fusion Protein Ag13915 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DXS253E P3 P3 protein SLC10A3; P3 protein; Protein P3; solute carrier family 10 (sodium/bile acid cotransporter family), member 3; Solute carrier family 10 member 3; solute carrier family 10, member 3
Gene Aliases: DXHXS253E; DXS253E; DXS253Eh; DXSmhG28; P3; SLC10A3
UniProt ID: (Human) P09131, (Mouse) P21129
Entrez Gene ID: (Human) 8273, (Mouse) 214601
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.