Novus Biologicals
Manufacturer Code:NBP188813
Catalog # NBP188813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:NKAEIPESKENGTEPESSFYKANGGFQPDE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apical sodium-dependent bile acid transporter; Apical sodium-dependent bile acid transporter ASBT IBAT ileal apical sodium-dependent bile acid transporter Ileal Na(+)/bile acid cotransporter ileal sodium/bile acid cotransporter Ileal sodium-dependent bile acid transporter ISBT Na(+)-dependent ileal bile acid transporter NTCP2PBAM Sodium/taurocholate cotransporting polypeptide ileal solute carrier family 10 (sodium/bile acid cotransporter family) member 2 Solute carrier family 10 member 2; ASBT; IBAT; ileal apical sodium-dependent bile acid transporter; Ileal Na(+)/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; Ileal sodium/bile acid cotransporter; Na(+)-dependent ileal bile acid transporter; Sodium/taurocholate cotransporting polypeptide, ileal; solute carrier family 10 (sodium/bile acid cotransporter family), member 2; solute carrier family 10 (sodium/bile acid cotransporter), member 2; Solute carrier family 10 member 2
Gene Aliases: ASBT; IBAT; ISBT; NTCP2; PBAM; SLC10A2
UniProt ID: (Human) Q12908
Entrez Gene ID: (Human) 6555
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.