Novus Biologicals
Manufacturer Code:NBP29318420UL
Catalog # NB028256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human SLC10A1 (NP_003040.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 29 protein; Cell growth-inhibiting gene 29 protein Na(+)/bile acid cotransporter Na(+)/taurocholate transport protein Na/taurocholate cotransporting polypeptide NTCP NTCP1 NTCPgrowth-inhibiting protein 29 sodium/bile acid cotransporter sodium/taurocholate cotransporter Sodium/taurocholate cotransporting polypeptide solute carrier family 10 (sodium/bile acid cotransporter family) member 1 Solute carrier family 10 member 1; growth-inhibiting protein 29; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; Na/taurocholate cotransporting polypeptide; Sodium/bile acid cotransporter; sodium/taurocholate cotransporter; Sodium/taurocholate cotransporting polypeptide; solute carrier family 10 (sodium/bile acid cotransporter family), member 1; solute carrier family 10 (sodium/bile acid cotransporter), member 1; Solute carrier family 10 member 1
Gene Aliases: GIG29; NTCP; SLC10A1
UniProt ID: (Human) Q14973
Entrez Gene ID: (Human) 6554
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.