Invitrogen
Manufacturer Code:PA541819
Catalog # PIPA541819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hairpin binding protein, histone; histone binding protein; Histone RNA hairpin-binding protein; histone stem loop binding protein; histone stem-loop binding protein; Histone stem-loop-binding protein; RNA binding protein; stem-loop (histone) binding protein; Stem-loop binding protein HBP
Gene Aliases: cb157; ele; HBP; sb:cb157; si:dkey-102m7.2; SLBP
UniProt ID: (Human) Q14493, (Mouse) P97440
Entrez Gene ID: (Dog) 488810, (Horse) 100052514, (Human) 7884, (Bovine) 444868, (Guinea pig) 100732116, (Mouse) 20492, (Rat) 681062, (Zebrafish) 407727
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.