Novus Biologicals
Manufacturer Code:NB10063446
Catalog # NB10063446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 9D1 |
Immunogen | CHO cells stably transfected with mouse CD150 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: CD 150 CD150IPO-3 CDw150 signaling lymphocytic activation molecule signaling lymphocytic activation molecule family member 1 SLAMCD150 antigen; CD150; CDw150; IPO-3; Signaling lymphocytic activation molecule; SLAM family member 1
Gene Aliases: CD150; CDw150; SLAM; SLAMF1
UniProt ID: (Human) Q13291
Entrez Gene ID: (Human) 6504
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.