Invitrogen
Manufacturer Code:PA532121
Catalog # PIPA532121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment contain a sequence corresponding to a region within amino acids 192 and 469 of SLA/LP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Liver-pancreas antigen; LP; LP PCH2D SLA SLA/LP O-phosphoseryl-tRNA(Sec) selenium transferase SLA-p35 SLA/LP autoantigen UGA suppressor tRNA-associated protein liver-pancreas antigen soluble liver antigen soluble liver antigen/liver pancreas antigen tRNA(Ser/Sec)-associated antigenic protein; O-phosphoseryl-tRNA(Sec) selenium transferase; Sec synthase; Selenocysteine synthase; Selenocysteinyl-tRNA(Sec) synthase; Sep-tRNA:Sec-tRNA synthase; SepSecS; SLA; SLA-p35; SLA/LP autoantigen; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; tRNA(Ser/Sec)-associated antigenic protein; UGA suppressor tRNA-associated protein
Gene Aliases: LP; PCH2D; SEPSECS; SLA; SLA/LP; TRNP48
UniProt ID: (Human) Q9HD40
Entrez Gene ID: (Human) 51091
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.