Proteintech
Manufacturer Code:115511AP150UL
Catalog # 50-172-7259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLA/LP Fusion Protein Ag2097 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Liver pancreas antigen LP Sec synthase Selenocysteine synthase Sep tRNA:Sec tRNA synthase SEPSECS SLA SLA p35 SLA/LP SLA/LP autoantigen Soluble liver antigen TRNP48; Liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; Sec synthase; Selenocysteine synthase; Selenocysteinyl-tRNA(Sec) synthase; Sep-tRNA:Sec-tRNA synthase; SepSecS; SLA; SLA-p35; SLA/LP autoantigen; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; tRNA(Ser/Sec)-associated antigenic protein; UGA suppressor tRNA-associated protein
Gene Aliases: 9130208G10; AA986712; D5Ertd135e; LP; PCH2D; SecS; SEPSECS; SLA; SLA/LP; TRNP48
UniProt ID: (Human) Q9HD40, (Mouse) Q6P6M7
Entrez Gene ID: (Human) 51091, (Rat) 679383, (Mouse) 211006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.