Proteintech
Manufacturer Code:127192AP150UL
Catalog # 50-172-8202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SKIV2L2 Fusion Protein Ag3414 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP dependent helicase SKIV2L2 Dob1 fSAP118 KIAA0052 Mtr4 SKIV2L2; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Exosome RNA helicase MTR4; functional spliceosome-associated protein 118; Superkiller viralicidic activity 2-like 2; TRAMP-like complex helicase
Gene Aliases: 2610528A15Rik; DOB1; fSAP118; KIAA0052; mKIAA0052; MTR4; MTREX; RGD1305984; SKIV2L2
UniProt ID: (Human) P42285, (Mouse) Q9CZU3
Entrez Gene ID: (Human) 23517, (Mouse) 72198, (Rat) 365668
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.