Novus Biologicals
Manufacturer Code:NBP247274
Catalog # NBP247274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LAELTKRLGALEEPDMTGQLVDLPEYYSWGEELTETQHMIQRRIMESVNGLKSLSAGRVVVVKNQEHHNALGVILQVSSNSTSRVFTTLVLCDKPLSQDPQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 170A DDX13helicase SKI2W EC 3.6.1 Helicase-like protein HLPEC 3.6.4.- SKI2 SKI2Wsuperkiller viralicidic activity 2 (S. cerevisiae homolog)-like SKIV2 superkiller viralicidic activity 2-like (S. cerevisiae) W; Helicase SKI2W; Helicase-like protein; HLP; Ski2; SKI2 homolog, superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like
Gene Aliases: 170A; DDX13; HLP; SKI2; SKI2W; SKIV2; SKIV2L; SKIV2L1; THES2; W
UniProt ID: (Human) Q15477
Entrez Gene ID: (Human) 6499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.