Proteintech
Manufacturer Code:192181AP150UL
Catalog # 50-173-2369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SKIL Fusion Protein Ag5282 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SKI like oncogene Ski like protein Ski related oncogene Ski related protein SKIL SNO SnoA SnoI SnoN; SKI-like oncogene; Ski-like protein; Ski-related oncogene; ski-related oncogene snoN; Ski-related protein; ski/sno related
Gene Aliases: SKIL; Skir; SNO; SnoA; SnoI; SnoN
UniProt ID: (Human) P12757, (Mouse) Q60665
Entrez Gene ID: (Human) 6498, (Rat) 114208, (Mouse) 20482
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.