Novus Biologicals
Manufacturer Code:NBP177305
Catalog # NBP177305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human Ski. The immunogen is located within the first 50 amino acids of Ski. Amino Acid Squence: CFQPHPGLQKTLEQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-ski Proto-oncogene c-Ski ski oncogene ski oncoprotein SKV v-ski avian sarcoma viral oncogene homolog v-ski sarcoma viral oncogene homolog (avian); Proto-oncogene c-Ski; Ski oncogene; ski oncoprotein; v-ski avian sarcoma viral oncogene homolog
Gene Aliases: SGS; SKI; SKV
UniProt ID: (Human) P12755
Entrez Gene ID: (Human) 6497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.