Novus Biologicals
Manufacturer Code:NBP172131
Catalog # NBP172131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid synthetic peptide near the C-terminus of human SKA1. The immunogen is located within amino acids 180 - 230 of SKA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C18orf24 chromosome 18 open reading frame 24 MGC10200 spindle and kinetochore associated complex subunit 1 spindle and kinetochore-associated protein 1 spindle and KT (kinetochore) associated 1; Spindle and kinetochore-associated protein 1; spindle and KT (kinetochore) associated 1
Gene Aliases: C18orf24; SKA1
UniProt ID: (Human) Q96BD8
Entrez Gene ID: (Human) 220134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.