Invitrogen
Manufacturer Code:PA575220
Catalog # PIPA575220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 201-250 of Human Six3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX3; Homeobox protein SIX3 Sine oculis homeobox homolog 3 SIX3 HPE2 Homeobox protein SIX6 Homeodomain protein OPTX2 Optic homeobox 2 SIX6 OPTX2 SIX9; Homeobox protein SIX6; Homeodomain protein OPTX2; Optic homeobox 2; Sine oculis homeobox homolog 3; Sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 3; sine oculis homeobox-like protein 6; sine oculis-related homeobox 3 homolog; sine oculis-related homeobox 6 homolog; Six9 protein
Gene Aliases: E130112M24Rik; HPE2; MCOPCT2; ODRMD; OPTX2; SIX3; Six3a; Six3alpha; Six3b; Six3beta; SIX6; SIX9
UniProt ID: (Human) O95343, (Human) O95475, (Mouse) Q62233, (Mouse) Q9QZ28
Entrez Gene ID: (Human) 6496, (Human) 4990, (Mouse) 20473, (Rat) 78974, (Mouse) 20476, (Rat) 314221
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.