Invitrogen
Manufacturer Code:PA595466
Catalog # PIPA595466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Six3 different from the related mouse sequence by two amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX3; Homeobox protein SIX3 Sine oculis homeobox homolog 3 SIX3; Sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3 homolog
Gene Aliases: E130112M24Rik; HPE2; SIX3; Six3a; Six3alpha; Six3b; Six3beta
UniProt ID: (Human) O95343, (Mouse) Q62233
Entrez Gene ID: (Human) 6496, (Mouse) 20473, (Rat) 78974
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.