Invitrogen
Manufacturer Code:PA535166
Catalog # PIPA535166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 138-166 amino acids from the central region of human SIX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX3; Sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; SIX3 Homeobox protein SIX3 Sine oculis homeobox homolog 3
Gene Aliases: HPE2; SIX3
UniProt ID: (Human) O95343
Entrez Gene ID: (Human) 6496
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.