Invitrogen
Manufacturer Code:PA541582
Catalog # PIPA541582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SIX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2
Gene Aliases: SIX2
UniProt ID: (Human) Q9NPC8, (Mouse) Q62232
Entrez Gene ID: (Horse) 100068793, (Rabbit) 100349565, (Human) 10736, (Dog) 481357, (Bovine) 539492, (Sheep) 101110829, (Mouse) 20472, (Rat) 366542, (Guinea pig) 100720717
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.