Novus Biologicals
Manufacturer Code:NBP29480120UL
Catalog # NB024113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human SIX2 (NP_058628.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX2; homeobox protein SIX2 sine oculis homeobox (Drosophila) homolog 2 Sine oculis homeobox homolog 2 sine oculis homeobox homolog 2 (Drosophila) SIX homeobox 2; Sine oculis homeobox homolog 2
Gene Aliases: SIX2
UniProt ID: (Human) Q9NPC8
Entrez Gene ID: (Human) 10736
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.