Proteintech
Manufacturer Code:115621AP150UL
Catalog # 50-559-228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SIX2 Fusion Protein Ag2124 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX2; Homeobox protein SIX2 Sine oculis homeobox homolog 2 SIX homeobox 2 SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2 homolog
Gene Aliases: SIX2
UniProt ID: (Human) Q9NPC8, (Mouse) Q62232
Entrez Gene ID: (Human) 10736, (Mouse) 20472, (Rat) 366542
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.