Invitrogen
Manufacturer Code:PA540532
Catalog # PIPA540532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SIX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein SIX1; Sine oculis homeobox homolog 1; sine oculis-related homeobox 1 homolog; SIX homeobox 1 BOS3 DFNA23 TIP39
Gene Aliases: BB138287; BOS3; DFNA23; SIX1; TIP39
UniProt ID: (Human) Q15475, (Mouse) Q62231
Entrez Gene ID: (Human) 6495, (Dog) 490719, (Rabbit) 100349440, (Sheep) 100381248, (Horse) 100060564, (Guinea pig) 100717285, (Mouse) 20471, (Rat) 114634
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.