Proteintech
Manufacturer Code:107091AP150UL
Catalog # 50-172-6530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SIX1 Fusion Protein Ag1044 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS3 DFNA23 Homeobox protein SIX1 Sine oculis homeobox homolog 1 SIX homeobox 1 SIX1 TIP39; Homeobox protein SIX1; Sine oculis homeobox homolog 1; sine oculis-related homeobox 1 homolog
Gene Aliases: BB138287; BOS3; DFNA23; SIX1; TIP39
UniProt ID: (Human) Q15475, (Mouse) Q62231
Entrez Gene ID: (Human) 6495, (Mouse) 20471
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.