Novus Biologicals
Manufacturer Code:NBP176267
Catalog # NBP176267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SIRT6. The immunogen is located within the last 50 amino acids of SIRT6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: EC 3.5.1.- NAD-dependent deacetylase sirtuin-6 SIR2L6sir2-related protein type 6 SIR2-like protein 6 sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae) sirtuin (silent mating type information regulation 2 S. cerevisiae homolog) 6 sirtuin 6 sirtuin type 6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2-like protein 6; sir2-related protein type 6; sirtuin type 6
Gene Aliases: SIR2L6; SIRT6
UniProt ID: (Human) Q8N6T7
Entrez Gene ID: (Human) 51548
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.