Abnova
Manufacturer Code:PAB12036
Catalog # 89-115-545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 300-355 of human SIRT6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of SIRT6.
For Research Use Only
Protein Aliases: NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2-like protein 6; sir2-related protein type 6; SIR2L6; sirtuin type 6
Gene Aliases: SIR2L6; SIRT6
UniProt ID: (Human) Q8N6T7
Entrez Gene ID: (Human) 51548
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.