Invitrogen
Manufacturer Code:MA524768
Catalog # PIMA524768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | Purified recombit human SIRT6 protein expressed in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810449N18RikAI043036Mono ADP ribosyltransferase sirtuin 6NAD-dependent protein deacetylase sirtuin-6Regulatory protein SIR2 homolog 6Regulatory protein SIR2 homologSIR2 like 6SIR2 like protein 6Sir2 related protein type 6SIR2-like protein 6SIR2; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2-like protein 6; sir2-related protein type 6; sirtuin type 6
Gene Aliases: 2810449N18Rik; AI043036; SIR2L6; SIRT6
UniProt ID: (Human) Q8N6T7, (Mouse) P59941
Entrez Gene ID: (Human) 51548, (Mouse) 50721, (Rat) 299638
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.