Invitrogen
Manufacturer Code:PA578447
Catalog # PIPA578447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SIRT4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NAD-dependent ADP-ribosyltransferase sirtuin-4; NAD-dependent protein deacetylase sirtuin-4; NAD-dependent protein lipoamidase sirtuin-4, mitochondrial; Regulatory protein SIR2 homolog 4; sir2-like 4; SIR2-like protein 4; SIR2L4 sirtuin 4; SIRT4; sirtuin type 4
Gene Aliases: SIR2L4; SIRT4
UniProt ID: (Human) Q9Y6E7
Entrez Gene ID: (Human) 23409
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.