Invitrogen
Manufacturer Code:PA568562
Catalog # PIPA568562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SIRT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NAD-dependent ADP-ribosyltransferase sirtuin-4; NAD-dependent deacetylase sirtuin-4; NAD-dependent protein deacetylase sirtuin-4; NAD-dependent protein lipoamidase sirtuin-4, mitochondrial; Regulatory protein SIR2 homolog 4; sir2-like 4; SIR2-like protein 4; SIRT4; SIRT4 SIR2L4 sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae) sirtuin silent mating type information regulation 2 homolog 4 (S. cerevisiae) MGC130046 MGC130047 MGC57437 SIR2L4 sirtuin 4 sir2-like 4 sirtuin 4 sirtuin type 4; sirtuin type 4
Gene Aliases: 4930596O17Rik; SIR2L4; SIRT4
UniProt ID: (Human) Q9Y6E7, (Bovine) Q1JQC6, (Mouse) Q8R216
Entrez Gene ID: (Pig) 100125349, (Human) 23409, (Horse) 100053520, (Sheep) 100217404, (Dog) 477507, (Bovine) 519328, (Rabbit) 100353055, (Mouse) 75387, (Rat) 304539, (Guinea pig) 100734585
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.