Proteintech
Manufacturer Code:664101IG150UL
Catalog # 50-173-6837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D8G10 |
Immunogen | SIRT2 Fusion Protein Ag7756 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5E5 antigen; mSIR2L2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sirtuin-2; Regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2 SIR2 like protein 2 SIR2L SIR2L2 SIRT2; SIR2-like protein 2; sir2-related protein type 2; sirtuin type 2; sirtuin-2
Gene Aliases: 5730427M03Rik; SIR2; SIR2L; SIR2L2; SIRT2
UniProt ID: (Human) Q8IXJ6, (Mouse) Q8VDQ8, (Rat) Q5RJQ4
Entrez Gene ID: (Human) 22933, (Pig) 100125964, (Mouse) 64383, (Rat) 361532
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.