Proteintech
Manufacturer Code:603031IG150UL
Catalog # 50-173-6396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7F6G6 |
Immunogen | SIRT1 Fusion Protein Ag17677 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 75SirT1; hSIR2; hSIR2 hSIRT1 SIR2 like protein 1 SIR2L1 SIRT1; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; Regulatory protein SIR2 homolog 1; Sir2; sir2-like 1; SIR2-like protein 1; SIR2alpha; SirtT1 75 kDa fragment; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1
Gene Aliases: AA673258; SIR2; Sir2a; SIR2alpha; SIR2L1; SIRT1
UniProt ID: (Human) Q96EB6, (Mouse) Q923E4
Entrez Gene ID: (Human) 23411, (Mouse) 93759
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.