Invitrogen
Manufacturer Code:MA530880
Catalog # PIMA530880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 32 |
Immunogen | Recombit Human SIRT1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 75SirT1; hSIR2; hSIRT1; NAD-dependent deacetylase sirtuin-1 OTTHUMP00000198111 OTTHUMP00000198112 SIR2-like protein 1 SIR2alpha hSIR2 hSIRT1 sir2-like 1 sirtuin type 1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; Regulatory protein SIR2 homolog 1; SIR2-like protein 1; SirtT1 75 kDa fragment; sirtuin type 1
Gene Aliases: SIR2; SIR2alpha; SIR2L1; SIRT1
UniProt ID: (Human) Q96EB6
Entrez Gene ID: (Human) 23411
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.