Invitrogen
Manufacturer Code:PA587383
Catalog # PIPA587383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SIRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75SirT1; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; Regulatory protein SIR2 homolog 1; Sir2; sir2-like 1; SIR2-like protein 1; SIR2alpha; SirtT1 75 kDa fragment; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1
Gene Aliases: AA673258; SIR2; Sir2a; SIR2alpha; SIR2L1; SIRT1
UniProt ID: (Human) Q96EB6, (Mouse) Q923E4
Entrez Gene ID: (Human) 23411, (Mouse) 93759, (Rat) 309757
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.