R&D Systems
Manufacturer Code:MAB4546SP
Catalog # MAB4546SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 602411 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human SIRP alpha/CD172a Gly27-Asn370 (predicted) Accession # P78324 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bit; Bit BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs Brain Ig-like molecule with tyrosine-based activation motifs CD172 antigen-like family member A CD172a CD172a antigen Inhibitory receptor SHPS-1 Macrophage fusion receptor MFR MFRtyrosine phosphatase SHP substrate 1 MYD1 MYD-1 MyD-1 antigen P84 protein tyrosine phosphatase non-receptor type substrate 1 PTPNS1 SHP substrate 1 SHPS1 SHPS-1 SHPS1CD172A signal-regulatory protein alpha Signal-regulatory protein alpha-1 Signal-regulatory protein alpha-2 Signal-regulatory protein alpha-3 SIRPA SIRPalpha SIRP-ALPHA-1 Sirp-alpha-2 SIRPalpha2 Sirp-alpha-3 SIRPtyrosine-protein phosphatase non-receptor type substrate 1; Brain Ig-like molecule with tyrosine-based activation motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172 antigen-like family member A; CD172a; Inhibitory receptor SHPS-1; Macrophage fusion receptor; MyD-1 antigen; p84; SHP substrate 1; signal-regulatory protein alpha; Signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; Sirp-alpha-1; Sirp-alpha-2; Sirp-alpha-3; tyrosine phosphatase SHP substrate 1; Tyrosine-protein phosphatase non-receptor type substrate 1
Gene Aliases: BIT; CD172A; MFR; MYD-1; MYD1; P84; PTPNS1; SHPS1; SIRP; SIRPA
UniProt ID: (Human) P78324
Entrez Gene ID: (Human) 140885
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.