Invitrogen
Manufacturer Code:PA529544
Catalog # PIPA529544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 84 and 356 of Human SIRP alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bit; BIT CD172A MFR MYD-1 P84 PTPNS1 SHPS1 SIRP CD172 antigen-like family member A brain-immunoglobulin-like molecule with tyrosine-based activation motifs inhibitory receptor SHPS-1 macrophage fusion receptor myd-1 antigen tyrosine phosphatase SHP substrate 1 tyrosine-protein phosphatase non-receptor; Brain Ig-like molecule with tyrosine-based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172 antigen-like family member A; CD172a; Inhibitory receptor SHPS-1; Macrophage fusion receptor; mSIRP-alpha1; MyD-1 antigen; p84; protein tyrosine phosphatase, non-receptor type substrate 1; SHP substrate 1; signal-regulatory protein alpha; Signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; Sirp-alpha-1; Sirp-alpha-2; Sirp-alpha-3; tyrosine phosphatase SHP substrate 1; Tyrosine-protein phosphatase non-receptor type substrate 1
Gene Aliases: AI835480; BIT; CD172A; MFR; MYD-1; MYD1; P84; PTPNS1; SHP-1; SHPS-1; SHPS1; SIRP; SIRPA
UniProt ID: (Human) P78324, (Mouse) P97797
Entrez Gene ID: (Human) 140885, (Mouse) 19261
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.