Novus Biologicals
Manufacturer Code:NBP220369
Catalog # NBP220369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SIPA1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase-activating protein Spa-1; GTPase-activating protein Spa-1 MGC102688 MGC17037 p130 SPA-1 signal-induced proliferation-associated 1 signal-induced proliferation-associated protein 1 sipa-1 SPA1signal-induced proliferation-associated gene 1; p130 SPA-1; signal-induced proliferation-associated gene 1; Signal-induced proliferation-associated protein 1; Sipa-1
Gene Aliases: SIPA1; SPA1
UniProt ID: (Human) Q96FS4
Entrez Gene ID: (Human) 6494
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.