Proteintech
Manufacturer Code:146381AP150UL
Catalog # 50-172-9607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SIN3A Fusion Protein Ag6219 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone deacetylase complex subunit Sin3a; KIAA0700 SIN3A; Paired amphipathic helix protein Sin3a; SIN3 homolog A, transcription regulator; SIN3 transcription regulator homolog A; SIN3A; transcriptional co-repressor Sin3A; Transcriptional corepressor Sin3a; transcriptional regulator, SIN3 yeast homolog A; transcriptional regulator, SIN3A
Gene Aliases: AW553200; Kiaa4126; mKIAA4126; mSin3A; Sin3; SIN3A
UniProt ID: (Human) Q96ST3, (Mouse) Q60520
Entrez Gene ID: (Human) 25942, (Rat) 363067, (Mouse) 20466
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
deacetylase
hydrolase
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.