Novus Biologicals
Manufacturer Code:NBP220363
Catalog # NBP220363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SIL1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAP; BAPMarinesco-Sjogren syndrome BiP-associated protein MSS nucleotide exchange factor SIL1 SIL1 homolog endoplasmic reticulum chaperone (S. cerevisiae) ULG5; BiP-associated protein; Nucleotide exchange factor SIL1; SIL1 homolog, endoplasmic reticulum chaperone; SIL1-like protein endoplasmic reticulum chaperone
Gene Aliases: BAP; MSS; SIL1; ULG5; UNQ545/PRO836
UniProt ID: (Human) Q9H173
Entrez Gene ID: (Human) 64374
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.