Abnova
Manufacturer Code:H00010280K
Catalog # 89-201-394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SIGMAR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SIGMAR1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Aging-associated gene 8 protein; FLJ25585 MGC3851 OPRS1 SR-BP1; SIG-1R; Sigma 1-type opioid receptor; Sigma non-opioid intracellular receptor 1; SR-BP; SR31747 binding protein 1; SR31747-binding protein
Gene Aliases: AAG8; ALS16; DSMA2; hSigmaR1; OPRS1; SIG-1R; sigma1R; SIGMAR1; SR-BP; SR-BP1; SRBP
UniProt ID: (Human) Q99720
Entrez Gene ID: (Human) 10280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.