Invitrogen
Manufacturer Code:PA5115900
Catalog # PIPA5115900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SIGLEC9 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD329; CD329 CDw329 FOAP-9 OBBP-LIKE protein FOAP-9 sialic acid binding Ig like lectin 9 sialic acid binding Ig-like lectin 9 Sialic acid-binding Ig-like lectin 9 Siglec SIGLEC9 siglec-9 UNQ668/PRO1302; CDw329; Protein FOAP-9; sialic acid binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin 9; Siglec-9
Gene Aliases: CD329; CDw329; FOAP-9; OBBP-LIKE; siglec-9; SIGLEC9; UNQ668/PRO1302
UniProt ID: (Human) Q9Y336
Entrez Gene ID: (Human) 27180
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.