Invitrogen
Manufacturer Code:MA524359
Catalog # PIMA524359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 750620 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Siglec-E Gln19-Phe354 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: MIS; Myeloid inhibitory siglec; S2V SIGLECL1 SLG Siglec-XII SIGLEC-like 1 sialic acid-binding Ig-like lectin 12 sialic acid-binding Ig-like lectin-like 1; sialic acid binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin E; Sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec-12; Siglec-5; Siglec-E; Siglec-L1; SIGLEC-like 1
Gene Aliases: Cd170; mSiglec-E; Siglec12; Siglec5; Siglec9; Siglece; Siglecl1
UniProt ID: (Mouse) Q91Y57
Entrez Gene ID: (Mouse) 83382
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.