Invitrogen
Manufacturer Code:PA5110369
Catalog # PIPA5110369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-210 of human SIGLEC12 (NP_443729.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cd170 MIS mSiglec-E Myeloid inhibitory siglec S2V Sialic acid-binding Ig-like lectin 12 Sialic acid-binding Ig-like lectin 5 Sialic acid-binding Ig-like lectin E Sialic acid-binding Ig-like lectin-like 1 Siglec Siglec12 Siglec-12 Siglec5 Siglec-5 Siglec9 Siglece Siglec-E Siglecl1 Siglec-L1 Siglec-XII SLG UNQ9215/PRO34042; MIS; Myeloid inhibitory siglec; sialic acid binding Ig-like lectin 12; sialic acid binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin E; Sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec-12; Siglec-5; Siglec-E; Siglec-L1; SIGLEC-like 1
Gene Aliases: Cd170; mSiglec-E; S2V; Siglec-XII; SIGLEC12; Siglec5; Siglec9; Siglece; SIGLECL1; SLG; UNQ9215/PRO34042
UniProt ID: (Human) Q96PQ1, (Mouse) Q91Y57
Entrez Gene ID: (Human) 89858, (Mouse) 83382
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule membrane-bound signaling molecule myelin protein receptor signaling molecule structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.