Abnova
Manufacturer Code:PAB6335
Catalog # 89-132-336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SIGLEC12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SIGLEC12.
For Research Use Only
Protein Aliases: FLJ38600; S2V; sialic acid binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin-like 1; Siglec-12; Siglec-L1; SIGLEC-like 1; Siglec-XII; SIGLECL1; SLG
Gene Aliases: S2V; Siglec-XII; SIGLEC12; SIGLECL1; SLG; UNQ9215/PRO34042
UniProt ID: (Human) Q96PQ1
Entrez Gene ID: (Human) 89858
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.