Invitrogen
Manufacturer Code:PA569064
Catalog # PIPA569064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human SIDT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SID1 transmembrane family member 2; SID1 transmembrane family, member 2; SIDT2 SID1 transmembrane family member 2 CGI-40
Gene Aliases: B930096O19; BC023957; CGI-40; PSEC0072; RGD1308311; SIDT2; UNQ685/PRO1325
UniProt ID: (Human) Q8NBJ9, (Mouse) Q8CIF6, (Rat) D3ZEH5
Entrez Gene ID: (Human) 51092, (Dog) 100856519, (Horse) 100062652, (Rabbit) 100346592, (Bovine) 513462, (Mouse) 214597, (Guinea pig) 100731389, (Rat) 315617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.