Novus Biologicals
Manufacturer Code:NBP227547
Catalog # NBP227547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RFQRKSIDGSFGSN corresponding to internal region according to NP_060169.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B830021E24Rik FLJ20174 SID1 SID-1 SID1 transmembrane family member 1 SID1 transmembrane family member 1; SID1 transmembrane family member 1; SID1 transmembrane family, member 1
Gene Aliases: SID-1; SID1; SIDT1
UniProt ID: (Human) Q9NXL6
Entrez Gene ID: (Human) 54847
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.