Proteintech
Manufacturer Code:553521AP150UL
Catalog # 50-173-6121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B830021E24Rik SID 1 SID1 SIDT1; SID1 transmembrane family member 1; SID1 transmembrane family, member 1
Gene Aliases: AW045928; B830021E24Rik; SID-1; SID1; SIDT1
UniProt ID: (Human) Q9NXL6, (Mouse) Q6AXF6
Entrez Gene ID: (Human) 54847, (Mouse) 320007
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.