Proteintech
Manufacturer Code:138861AP150UL
Catalog # 50-172-9066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SIAH1 Fusion Protein Ag4336 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase SIAH1; E3 ubiquitin-protein ligase SIAH1A; hSIAH1 HUMSIAH Seven in absentia homolog 1 Siah 1 Siah 1a SIAH1; mSiah-1a; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH1A; Seven in absentia homolog 1; Seven in absentia homolog 1a; Siah-1; Siah-1a
Gene Aliases: AA982064; AI853500; HUMSIAH; SIAH1; SIAH1A; Sinh1a
UniProt ID: (Human) Q8IUQ4, (Mouse) P61092
Entrez Gene ID: (Human) 6477, (Mouse) 20437
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.